EMAIL THIS PAGE TO A FRIEND
Acta cirurgica brasileira
Erdosteine ameliorates the harmful effects of ischemia-reperfusion injury on the liver of rats.
PMID 29160366
Abstract
To investigate the potential protective effects of erdosteine against the harmful effects of ischemia-reperfusion injury on the liver in an experimental rat model. Forty rats were divided into 4 groups. In the sham group, only the hepatic pedicle was mobilized. No other manipulation or treatment was performed. In the other groups, ischemia was achieved by clamping the hepatic pedicle for 60 min. After that, 90 min reperfusion was provided. In the control group, no treatment was given. In the perioperative treatment group, 100 mg/kg erdosteine was administered 2 hours before ischemia induction. In the preoperative treatment group, 100 mg/kg/day erdosteine was administered daily for ten days before the operation. At the end of the procedures, blood and liver samples were obtained for biochemical and histopathological assessment. Treatment with erdosteine ameliorated the histopathological abnormalities when compared with the control group. Furthermore, this treatment significantly decreased the serum liver function test values. It was also found that erdosteine ameliorated the oxidative stress parameters in both the perioperative and preoperative treatment groups. The current study is the first to have shown the favorable effects of erdosteine on the harmful effects of experimental hepatic ischemia-reperfusion injury.