EMAIL THIS PAGE TO A FRIEND
Scientific reports
Silencing of OsGRXS17 in rice improves drought stress tolerance by modulating ROS accumulation and stomatal closure.
PMID 29162892
Abstract
Glutaredoxins (GRXs) modulate redox-dependent signaling pathways and have emerged as key mediators in plant responses to environmental stimuli. Here we report that RNAi-mediated suppression of Oryza sativa GRXS17 (OsGRXS17) improved drought tolerance in rice. Gene expression studies showed that OsGRXS17 was present throughout the plant and that transcript abundance increased in response to drought stress and abscisic acid (ABA) treatment. Localization studies, utilizing GFP-OsGRXS17 fusion proteins, indicated that OsGRXS17 resides in both the cytoplasm and the nuclear envelope. Under drought stress conditions, rice plants with reduced OsGRXS17 expression showed lower rates of water loss and stomatal conductance, higher relative water content, and enhanced survival compared to wild-type controls. Further characterization of the OsGRXS17 down-regulated plants revealed an elevation in H