EMAIL THIS PAGE TO A FRIEND
Frontiers in behavioral neuroscience
Behavioral and Neurochemical Phenotyping of Mice Incapable of Homer1a Induction.
PMID 29163080
Abstract
Immediate early and constitutively expressed products of the