EMAIL THIS PAGE TO A FRIEND
Oncotarget
Myeloid ecotropic viral integration site 1 inhibits cell proliferation, invasion or migration in human gastric cancer.
PMID 29163810
Abstract
Myeloid ecotropic viral integration site 1 (MEIS1) has been identified to be a potential tumor suppressor in some cancers. However, the mechanisms underlying MEIS1-induced cancer development and progression were not clear. Here, we investigated the expression and role of MEIS1 in gastric cancer.