EMAIL THIS PAGE TO A FRIEND
Journal of immunology (Baltimore, Md. : 1950)
Route of Antigen Presentation Can Determine the Selection of Foxp3-Dependent or Foxp3-Independent Dominant Immune Tolerance.
PMID 29167234
Abstract
It has been shown that dominant tolerance, namely in transplantation, requires Foxp3