EMAIL THIS PAGE TO A FRIEND
The Journal of neuroscience : the official journal of the Society for Neuroscience
Tonotopic Variation of the T-Type Ca
PMID 29167400
Abstract
Neurons in avian nucleus laminaris (NL) are binaural coincidence detectors for sound localization and are characterized by striking structural variations in dendrites and axon initial segment (AIS) according to their acoustic tuning [characteristic frequency (CF)]. T-type Ca