EMAIL THIS PAGE TO A FRIEND
Frontiers in pharmacology
Volatile Oil from Amomi Fructus Attenuates 5-Fluorouracil-Induced Intestinal Mucositis.
PMID 29170638
Abstract
Amomi Fructus has been used to treat digestive diseases in the context of traditional Chinese medicine, so we evaluated the effects of a volatile oil from