EMAIL THIS PAGE TO A FRIEND
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association
Aflatoxin exposure in Nigerian children with severe acute malnutrition.
PMID 29175577
Abstract
Aflatoxin exposure is an important public health concern in sub-Saharan Africa as well as parts of Latin America and Asia. In addition to hepatocellular carcinoma, chronic aflatoxin exposure is believed to play a role in childhood growth impairment. The most reliable biomarker of chronic aflatoxin exposure is the aflatoxin-albumin adduct, as measured by ELISA or isotope dilution mass spectrometry (IDMS). In this report, we have used high resolution LC-MS/MS with IDMS to quantitate AFB
Related Materials