EMAIL THIS PAGE TO A FRIEND
Oncotarget
SH003 suppresses breast cancer growth by accumulating p62 in autolysosomes.
PMID 29179443
Abstract
Drug markets revisits herbal medicines, as historical usages address their therapeutic efficacies with less adverse effects. Moreover, herbal medicines save both cost and time in development. SH003, a modified version of traditional herbal medicine extracted from