EMAIL THIS PAGE TO A FRIEND
Oncotarget
Sphingosine kinase 1 mediates diabetic renal fibrosis via NF-κB signaling pathway: involvement of CK2α.
PMID 29179493
Abstract
Sphingosine kinase 1 (SphK1) plays a pivotal role in regulating diabetic renal fibrotic factors such as fibronectin (FN) and intercellular adhesion molecule-1 (ICAM-1). Especially, activation of SphK1 is closely linked to the body inflammatory reaction. Casein kinase 2α subunit (CK2α), a protein kinase related to inflammatory reaction, influences diabetic renal fibrosis and expressions of FN and ICAM-1 via NF-κB pathway. However, the mechanism by which SphK1 mediates diabetic renal fibrosis has not yet fully elucidated. The current study is aimed to investigate if SphK1 mediates diabetic renal fibrotic pathological process via inflammatory pathway and activation of CK2α. The following findings were observed: (1) Expressions of SphK1 were upregulated in kidneys of diabetic mice and rats; (2) Knockdown of SphK1 expression suppressed high glucose (HG)-induced NF-κB nuclear translocation and expressions of FN and ICAM-1; (3) Compared with C57 diabetic mice, SphK1