EMAIL THIS PAGE TO A FRIEND
Beilstein journal of nanotechnology
Surfactant-induced enhancement of droplet adhesion in superhydrophobic soybean (
PMID 29181291
Abstract
This study performed with soybean (