EMAIL THIS PAGE TO A FRIEND
Experimental cell research
miR-195 inhibited abnormal activation of osteoblast differentiation in MC3T3-E1 cells via targeting RAF-1.
PMID 29197556
Abstract
Recent reports have demonstrated that RAF-1 To demonstrate that RAF-1 is associated with bone deformity and that RAF-1 RAF-1, especially RAF-1 Our findings indicated that miR-195 inhibited WT and L613V RAF-1 induced hyperactive osteoblast differentiation in MC3T3-E1 cells by targeting RAF-1. miR-195 might be a novel therapeutic agent for the treatment of L613V-induced bone deformity in Noonan syndrome.