EMAIL THIS PAGE TO A FRIEND
Cell systems
Linear Integration of ERK Activity Predominates over Persistence Detection in Fra-1 Regulation.
PMID 29199017
Abstract
ERK signaling regulates the expression of target genes, but it is unclear how ERK activity dynamics are interpreted. Here, we investigate this question using simultaneous, live, single-cell imaging of two ERK activity reporters and expression of Fra-1, a target gene controlling epithelial cell identity. We find that Fra-1 is expressed in proportion to the amplitude and duration of ERK activity. In contrast to previous "persistence detector" and "selective filter" models in which Fra-1 expression only occurs when ERK activity persists beyond a threshold duration, our observations demonstrate that the network regulating Fra-1 expression integrates total ERK activity and responds to it linearly. However, exploration of a generalized mathematical model of the Fra-1 coherent feedforward loop demonstrates that it can perform either linear integration or persistence detection, depending on the basal mRNA production rate and protein production delays. Our data indicate that significant basal expression and short delays cause Fra-1 to respond linearly to integrated ERK activity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C8052 Cholera Toxin from Vibrio cholerae, ~95% (SDS-PAGE), lyophilized powder