EMAIL THIS PAGE TO A FRIEND
Experimental and therapeutic medicine
Leptin upregulates COX-2 and its downstream products in aortic endothelial cells.
PMID 29201221
Abstract
The adipocyte-derived hormone leptin is associated with hypertension. The involvement of cyclooxygenase-2 (COX-2) and its downstream vasomotor products prostaglandin (PG) and thromboxane (TX)A