EMAIL THIS PAGE TO A FRIEND
Antimicrobial agents and chemotherapy
Chemogenomic Profiling of the Fungal Pathogen Candida albicans.
PMID 29203491
Abstract
There is currently a small number of classes of antifungal drugs, and these drugs are known to target a very limited set of cellular functions. We derived a set of approximately 900 nonessential, transactivator-defective disruption strains from the tetracycline-regulated GRACE collection of strains of the fungal pathogen
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
32103
Posaconazole, VETRANAL, analytical standard
C37H42F2N8O4