EMAIL THIS PAGE TO A FRIEND
Oncotarget
Immunization with recombinant rabies virus expressing Interleukin-18 exhibits enhanced immunogenicity and protection in mice.
PMID 29207661
Abstract
Several studies have shown that interleukin-18 (IL-18) plays an important role in both innate and adaptive immune responses. In this study, we investigated the pathogenicity and immunogenicity of recombinant rabies virus expressing IL-18 (rHEP-IL18). Experimental results showed that Institute of Cancer Research (ICR) mice that received a single intramuscular immunization with rHEP-IL18 elicited the highest titers of serum neutralizing antibodies and the strongest cell-mediated immune responses to prevent the development of rabies disease, compared with immunization with the parent virus HEP-Flury. Mice inoculated with rHEP-IL18 developed significantly higher IFN-γ responses, increased percentages of CD4