EMAIL THIS PAGE TO A FRIEND
European journal of pharmacology
Role of transglutaminase 2 in A
PMID 29208472
Abstract
Pharmacologically-induced pre- and post-conditioning represent attractive therapeutic strategies to reduce ischaemia/reperfusion injury during cardiac surgery and following myocardial infarction. We have previously reported that transglutaminase 2 (TG2) activity is modulated by the A