EMAIL THIS PAGE TO A FRIEND
Oncotarget
Silencing of lncRNA AFAP1-AS1 suppressed lung cancer development by regulatory mechanism in
PMID 29212176
Abstract
Although the long noncoding RNA AFAP1-AS1 has been shown to be involved in various types of cancer, its involvement in lung cancer remains poorly understood. In the current study, we found that AFAP1-AS1 was substantially over expressed in lung cancer tissues and cell lines. In addition, AFAP1-AS1 expression level was proven to be associated with the malignant features of lung cancer. Knockdown of AFAP1-AS1 significantly suppressed cell proliferation by increasing cell apoptosis and G0/G1 phase retardation of cell cycle in lung cancer cells. Furthermore, AFAP1-AS1 knockdown could suppress tumor growth of lung cancer in BALB/c nude mice. We also identified that AFAP1-AS1 silencing could influence the expression of AFAP1 and KRT1 on mRNA and protein level by