EMAIL THIS PAGE TO A FRIEND
Oncotarget
Insulin-like growth factor 2 mRNA binding protein 3 (IGF2BP3) promotes lung tumorigenesis via attenuating p53 stability.
PMID 29212181
Abstract
Insulin-like growth factor 2 mRNA binding protein 3 (IGF2BP3/IMP3/KOC), initially identified as an RNA-binding protein, is highly expressed in embryonic tissues and a variety of cancers. Previously, our group reported that IGF2BP3 may serve as a potential diagnostic marker for lung cancer. However, little is known about the function of IGF2BP3 in lung cancer development. Here we demonstrate that IGF2BP3 expression was markedly increased in lung cancer tissues compared to normal tissues at both mRNA and protein levels. Overexpression of IGF2BP3 in lung cancer cells promoted cell proliferation, tumor migration and invasion