EMAIL THIS PAGE TO A FRIEND
Oncotarget
Plasma-derived extracellular vesicle proteins as a source of biomarkers for lung adenocarcinoma.
PMID 29221141
Abstract
Exosomes and other extracellular vesicles (EVs) have been implicated as mediators of intercellular communication. Their release into the circulation has the potential to inform about tumor status. In-depth proteomic characterization of plasma-derived EVs has been limited by challenges in isolating EVs from protein-abundant biological fluids. We implemented a novel single-step density gradient flotation workflow for efficient and rapid isolation of highly enriched circulating EVs from plasma. Mass-spectrometry analysis of plasma EVs from subjects with lung adenocarcinoma and matched controls resulted in the identification of 640 proteins. A total of 108 proteins exhibited significant (