EMAIL THIS PAGE TO A FRIEND
Oncotarget
Direct HPV E6/Myc interactions induce histone modifications, Pol II phosphorylation, and hTERT promoter activation.
PMID 29221209
Abstract
Human Papillomavirus Viruses (HPVs) are associated with the majority of human cervical and anal cancers and 10-30% of head and neck squamous carcinomas. E6 oncoprotein from high risk HPVs interacts with the p53 tumor suppressor protein to facilitate its degradation and increases telomerase activity for extending the life span of host cells. We published previously that the Myc cellular transcription factor associates with the high-risk HPV E6 protein