EMAIL THIS PAGE TO A FRIEND
Journal of agricultural and food chemistry
Isobaric Tags for Relative and Absolute Quantification (iTRAQ)-Based Untargeted Quantitative Proteomic Approach To Identify Change of the Plasma Proteins by Salbutamol Abuse in Beef Cattle.
PMID 29240408
Abstract
Salbutamol, a selective β