EMAIL THIS PAGE TO A FRIEND
Oncotarget
Identification of glycerol-3-phosphate dehydrogenase 1 as a tumour suppressor in human breast cancer.
PMID 29254166
Abstract
In the present study, we found the mRNA expression level of glycerol-3-phosphate dehydrogenase (GPD1) was significantly downregulated in human breast cancer patients. Patients with reduced GPD1 expression exhibited poorer overall metastatic relapse-free survival (