EMAIL THIS PAGE TO A FRIEND
Frontiers in chemistry
Redox-Responsive Manganese Dioxide Nanoparticles for Enhanced MR Imaging and Radiotherapy of Lung Cancer.
PMID 29255705
Abstract
In this study, we synthesized manganese dioxide nanoparticles (MnO