EMAIL THIS PAGE TO A FRIEND
Frontiers in plant science
A Novel AP2/ERF Transcription Factor CR1 Regulates the Accumulation of Vindoline and Serpentine in
PMID 29270185
Abstract
As one type of the most important alkaloids in the world, terpenoid indole alkaloids (TIAs) show a wide range of pharmaceutical activities that are beneficial for clinical treatments.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
14764
1-Deoxy-D-xylulose, ≥80% (TLC)
C5H10O4