EMAIL THIS PAGE TO A FRIEND
Dalton transactions (Cambridge, England : 2003)
Photocatalytic function of the B
PMID 29271445
Abstract
A visible light induced three-component catalytic system with the cobalamin derivative (B
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
135542
Diethyl benzylmalonate, 98%
C14H18O4