EMAIL THIS PAGE TO A FRIEND
Frontiers in microbiology
Synergistic Antimicrobial Interaction between Honey and Phage against
PMID 29276503
Abstract
Chronic wounds afford a hostile environment of damaged tissues that allow bacterial proliferation and further wound colonization.