EMAIL THIS PAGE TO A FRIEND
Carbohydrate polymers
Novel cationic chitosan derivative bearing 1,2,3-triazolium and pyridinium: Synthesis, characterization, and antifungal property.
PMID 29279113
Abstract
In this paper, novel cationic chitosan derivative possessing 1,2,3-triazolium and pyridinium groups was synthesized conveniently via cuprous-catalyzed azide-alkyne cycloaddition (CuAAC) and methylation. FTIR,
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A78209
3-Aminopyridine, 99%
C5H6N2