EMAIL THIS PAGE TO A FRIEND
Oncotarget
Nuclear membrane-localised NOX4D generates pro-survival ROS in FLT3-ITD-expressing AML.
PMID 29285262
Abstract
Internal tandem duplication of the juxtamembrane domain of FMS-like tyrosine kinase 3 (FLT3-ITD) is the most prevalent genetic aberration present in 20-30% of acute myeloid leukaemia (AML) cases and is associated with a poor prognosis. FLT3-ITD expressing cells express elevated levels of NADPH oxidase 4 (NOX4)-generated pro-survival hydrogen peroxide (H