EMAIL THIS PAGE TO A FRIEND
Oncotarget
Targeting G-protein coupled receptor-related signaling pathway in a murine xenograft model of appendiceal pseudomyxoma peritonei.
PMID 29290997
Abstract
Cancer cells aberrantly express mucins to enhance their survival. Relative chemoresistance of appendiceal pseudomyxoma peritonei (PMP) is attributed to abundant extracellular mucin 2 (MUC2) protein production. We hypothesized that simultaneous MUC2 inhibition and apoptosis induction would be effective against mucinous tumors.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
EHU092951 MISSION® esiRNA, esiRNA human CREB1 (esiRNA1)