EMAIL THIS PAGE TO A FRIEND
Nature communications
Macrophages orchestrate breast cancer early dissemination and metastasis.
PMID 29295986
Abstract
Cancer cell dissemination during very early stages of breast cancer proceeds through poorly understood mechanisms. Here we show, in a mouse model of HER2