EMAIL THIS PAGE TO A FRIEND
Biochimica et biophysica acta
The primary structure of human apolipoprotein A-IV.
PMID 2930771
Abstract
Human apolipoprotein (apo) A-IV was purified from chylous ascites fluid. Proteolytic peptides produced by trypsin and Staphylococcus aureus V8 proteinase digestions were purified by high-performance liquid chromatography and sequenced. Human apoA-IV contains 376 amino acid residues. The peptide-derived sequence generally matches two previously reported DNA-derived amino acid sequences except for discrepancies in five positions. In order to examine these discrepancies further, one complete apoA-IV cDNA clone and another partial clone were sequenced. Comparison of all the available information indicates that the peptide-derived sequence reported here is accurate. Sequencing errors probably account for some of the discrepancies between the two primary sequences predicted by earlier nucleotide analyses. In certain positions, however, bona fide sequence heterogeneity or cloning artifact cannot be excluded.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AV54316
Anti-APOA4 antibody produced in rabbit, affinity isolated antibody