EMAIL THIS PAGE TO A FRIEND
Journal of fluorine chemistry
Computational Design and Synthesis of Novel Fluoro-Analogs of Combretastatins A-4 and A-1.
PMID 29311752
Abstract
Combretastatin A-1 (CA-1) and combretastatin A-4 (CA-4) isolated from the African bush willow