EMAIL THIS PAGE TO A FRIEND
Frontiers in pharmacology
Dihydromyricetin Protects against Bone Loss in Ovariectomized Mice by Suppressing Osteoclast Activity.
PMID 29311931
Abstract
Dihydromyricetin (DMY), the main flavonoid component of