EMAIL THIS PAGE TO A FRIEND
Contact dermatitis
Contact dermatitis to Butin-2-diol 1,4.
PMID 2931240
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B103209
2-Butyne-1,4-diol, 99%
C4H6O2