EMAIL THIS PAGE TO A FRIEND
American Journal of Cancer Research
SP1-induced up-regulation of lncRNA SNHG14 as a ceRNA promotes migration and invasion of clear cell renal cell carcinoma by regulating N-WASP.
PMID 29312804