EMAIL THIS PAGE TO A FRIEND
Microbiological reviews
Gene organization and regulation in the qa (quinic acid) gene cluster of Neurospora crassa.
PMID 2931582
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
46944-U
(−) Quinic acid, analytical standard
C7H12O6
138622
D-(−)-Quinic acid, 98%
C7H12O6