EMAIL THIS PAGE TO A FRIEND
Food chemistry
A survey of free glutamic acid in foods using a robust LC-MS/MS method.
PMID 29329874
Abstract
An effective and simultaneous liquid chromatography-tandem mass spectrometry (LC-MS/MS) method was used with the aim of quantifying monosodium glutamate (MSG) in foodstuffs, such as chips, taste cubes, sauces and soups. The results were linear (R
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
249467
Propyl chloroformate, 98%
C4H7ClO2