EMAIL THIS PAGE TO A FRIEND
Proceedings of the National Academy of Sciences of the United States of America
Infected erythrocytes expressing DC13 PfEMP1 differ from recombinant proteins in EPCR-binding function.
PMID 29339517
Abstract
Recent advances have identified a new paradigm for cerebral malaria pathogenesis in which endothelial protein C receptor (EPCR) is a major host receptor for sequestration of