EMAIL THIS PAGE TO A FRIEND
Zhong yao tong bao (Beijing, China : 1981)
[Constituents of the Lipophilic fraction of the Apocynum venetum leaf].
PMID 2934156
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
74156
Nonacosane, analytical standard
C29H60
284246
Nonacosane, 99%
C29H60