EMAIL THIS PAGE TO A FRIEND
Oncotarget
Inactivation/deficiency of DHODH induces cell cycle arrest and programed cell death in melanoma.
PMID 29348830
Abstract
Malignant melanoma (MM) is one of the most malignant tumors and has a very poor prognosis. However, there are no effective drugs to treat this disease. As a kind of iron flavin dependent enzyme, dihydroorotate dehydrogenase (DHODH, EC 1.3.3.1) is the fourth and a key enzyme in the