EMAIL THIS PAGE TO A FRIEND
Analytical and bioanalytical chemistry
A sensitive method using SPME pre-concentration for the quantification of aromatic amines in indoor air.
PMID 29353429
Abstract
Monitoring the levels of aliphatic and aromatic amines (AA) in indoor air is important to protect human health because of exposure to these compounds through diet and inhalation. A sampling and analytical method using XAD-2 cartridges and gas chromatography coupled to mass spectrometry used for assessing 25 AA in different smoking and non-smoking indoor environment was developed. After sampling and delivering 1 m
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
24241-U Carboxen®-1006 PLOT Capillary GC Column, L × I.D. 30 m × 0.32 mm, average thickness 15 μm
25461 Carboxen®-1006 PLOT Capillary GC Column, L × I.D. 30 m × 0.53 mm, average thickness 30 μm
24246 Carboxen®-1010 PLOT Capillary GC Column, L × I.D. 30 m × 0.32 mm, average thickness 15 μm
25467 Carboxen®-1010 PLOT Capillary GC Column, L × I.D. 30 m × 0.53 mm, average thickness 30 μm
54041-U Carboxen® 1000 Reversible SPE Tube, 200 mg/mL, pk 30
29753-U Carboxen® 569, O.D. × L 1/4  in. × 3 1/2 , glass TD tube, fritted, Sealed with Brass Endcaps, preconditioned, pkg of 10 ea
29534-U Carboxen® 569, glass TD tube, fritted, O.D. × L 1/4  in. × 3 1/2  in., preconditioned, pkg of 10 ea
54293-U Carboxen® 572 for Tobacco Smoke, Adsorbent Bed Wt: (300 mg), cartridge volume 3 mL, pkg of 10 ea
54294-U Carboxen® 572 for Tobacco Smoke, Adsorbent Bed Wt: 300 mg, cartridge size 3 mL, pkg of 50 ea
10263 Carboxen® Adsorbent, matrix Carboxen® 563, 20-45 mesh, bottle of 10 g
10264 Carboxen® Adsorbent, matrix Carboxen® 564, 20-45 mesh, bottle of 10 g
11324-U Carboxen® Adsorbent, matrix Carboxen® 564, 20-45 mesh, pack of 144 × 290 mg
10269 Carboxen® Adsorbent, matrix Carboxen® 569, 20-45 mesh, bottle of 10 g
11048-U Carboxen® Adsorbent, matrix Carboxen® 569, 20-45 mesh, bottle of 500 g
11072-U Carboxen® Adsorbent, matrix Carboxen® 572, 20-45 mesh, bottle of 10 g
10477-U Carboxen® Adsorbent, matrix Carboxen® 1000, 40-60 mesh, bottle of 50 g
10478-U Carboxen® Adsorbent, matrix Carboxen® 1000, 60-80 mesh, bottle of 10 g
10471 Carboxen® Adsorbent, matrix Carboxen® 1003, 40-60 mesh, bottle of 10 g
23446-U ORBO-93 Sorbent Tube, matrix Carboxen® 1000, ORBO tube I.D. × L 6 mm × 95 mm, Bed B 90 mg, Bed A 180 mg, 60/80 mesh, pkg of 25 ea
20355 ORBO 78 HBr on Carboxen® 564 specially cleaned (20/45), 400/200 mg, W,W,W separators, O.D. × L 6 mm × 110 mm, pkg of 25 ea
20358 ORBO 90 Carboxen® 564 (20/45), 160/80 mg, W,F,F separators, O.D. × L 6 mm × 75 mm, pkg of 25 ea, for analyte group MEK (methylethyl ketone)
28718-U Tenax® TA / Carboxen® 1018, glass TD tube, preconditioned, O.D. × L 1/4  in. × 3 1/2  in., Sealed with (Swagelok® End-Fittings), pkg of 10 ea