EMAIL THIS PAGE TO A FRIEND
Biotechnology & genetic engineering reviews
Low-molecular-weight immunomodifiers produced by micro-organisms.
PMID 2936359
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E0761
Ebelactone A microbial, esterase inhibitor
C20H34O4