EMAIL THIS PAGE TO A FRIEND
Organic & biomolecular chemistry
DBU-promoted carbonylative synthesis of 1,3-oxathiolan-2-ones from propargylic alcohols with TFBen as the CO source.
PMID 29363708
Abstract
A DBU-promoted carbonylative cyclization of propargylic alcohols with sulfur was developed. Various 1,3-oxathiolan-2-ones were produced in 61-98% yields under mild conditions in the absence of metal catalysts. TFBen (benzene-1,3,5-triyl triformate) as an efficient and solid CO surrogate and S
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
901821
Benzene-1,3,5-triyl triformate
C9H6O6