EMAIL THIS PAGE TO A FRIEND
Anticancer research
Inhibition of Sirtuin 6 Induces Neuroblastoma Differentiation.
PMID 29374686
Abstract
Sirtuins (SIRTs) play crucial roles in various signaling pathways that modulate differentiation and proliferation. We sought to elucidate the role of SIRTs in differentiation and proliferation of human neuroblastoma (NB). NB cells were treated with nicotinamide (NAM), a non-specific SIRT inhibitor, SIRT-targeted short hairpin RNAs, and retinoic acid to assess cell growth and differentiation. SIRTs are involved in proliferation and differentiation using NAM in BE(2)-C cells. Specifically, SIRT6 knockdown in BE(2)-C cells reduced cell proliferation, induced neurite extension, corresponding with induction of p21 SIRTs have important oncogenic properties in NB beyond its established functions in aging and genome stability. SIRT6 may represent a novel target for developing future therapeutics for the treatment of aggressive NBs.