EMAIL THIS PAGE TO A FRIEND
Ceskoslovenska epidemiologie, mikrobiologie, imunologie
[Additional comments on the possibilities of using Chloramine B and Dikonit in a mixture with detergents].
PMID 2937549
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
23265
N-Chlorobenzenesulfonamide sodium salt, ~28% active chlorine basis
C6H5ClNNaO2S · xH2O