EMAIL THIS PAGE TO A FRIEND
Clinical chemistry
Direct determination of sulfate conjugates of 17-oxosteroids in urine by liquid chromatography.
PMID 2938850
Abstract
We describe a new liquid-chromatographic method with fluorescence detection for direct simultaneous determination of four urinary sulfate conjugates of 17-oxosteroids (17OS), without use of solvolysis or enzymatic hydrolysis. The 17OS sulfates in urine samples are extracted with benzene as ion pairs in the presence of benzyltributylammonium chloride. These ion pairs are converted to hydrazones by use of dansyl hydrazine in trichloroacetic acid solution and then are separated by liquid chromatography. The proposed method, being simple and rapid, shows a good separation of 17OS sulfates and superior sensitivity, sufficient recovery, and good reproducibility.