EMAIL THIS PAGE TO A FRIEND
Cells, tissues, organs
Porous Chitosan/Nano-Hydroxyapatite Composite Scaffolds Incorporating Simvastatin-Loaded PLGA Microspheres for Bone Repair.
PMID 29393155
Abstract
The combination of bone tissue scaffolds with osteogenic induction factors is an effective strategy to facilitate bone healing processes. Here, chitosan (CS)/nano-hydroxyapatite (HA) scaffolds containing simvastatin (SIM)-loaded PLGA microspheres were fabricated by combining a freeze-drying technique with a modified water-oil-water emulsion method. The CS/HA weight ratio of 1:2 was selected by analyzing the effect of HA content on the micro-architecture, mechanical strength, and biocompatibility of the scaffold. Drug release kinetics showed that the SIM encapsulated in the scaffold was released in a sustained manner for up to 30 days. In vitro bioactivity study in rat bone marrow-derived mesenchymal stem cells showed that the SIM-loaded scaffolds had a strong ability in accelerating cell proliferation and inducing osteogenic differentiation. Moreover, an in vivo experiment using a rat calvarial defect model also documented that the SIM-loaded scaffolds had a remarkable effect on bone-promoting regeneration. The results of this study suggest that the SIM-loaded CS/HA scaffold is feasible and effective in bone repair and thus may provide a promising route for the treatment of critical-sized bone defects.