EMAIL THIS PAGE TO A FRIEND
Frontiers in physiology
Pretreatment with AQP4 and NKCC1 Inhibitors Concurrently Attenuated Spinal Cord Edema and Tissue Damage after Spinal Cord Injury in Rats.
PMID 29403391
Abstract
Spinal cord injury (SCI) affects more than 2.5 million people worldwide. Spinal cord edema plays critical roles in the pathological progression of SCI. This study aimed to delineate the roles of aquaporin 4 (AQP4) and Na
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0136
TGN-020, ≥98% (HPLC)
C8H6N4OS