EMAIL THIS PAGE TO A FRIEND
Clinics in dermatology
Dermatitis-producing Anacardiaceae of the Caribbean area.
PMID 2941128
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
91822
5-Pentadecylresorcinol, analytical standard
C21H36O2